• USA Home
  • A4177 - n-Amyl β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
A4177 Sigma-Aldrich

n-Amyl β-D-glucopyranoside

~98%

Synonym: n-Pentyl β-D-glucopyranoside

  • CAS Number 66957-71-9

  • Empirical Formula (Hill Notation) C11H22O6

  • Molecular Weight 250.29

  •  MDL number MFCD00057506

Purchase

Properties

assay   ~98%
storage temp.   −20°C
SMILES string   CCCCCOC1OC(CO)C(O)C(O)C1O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?