• USA Home
  • A4309 - [Gln11]-Amyloid β-Protein fragment 1-16

EMAIL THIS PAGE TO A FRIEND
A4309 Sigma-Aldrich

[Gln11]-Amyloid β-Protein fragment 1-16

≥97% (HPLC)

Synonym: Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Gln-Val-His-His-Gln-Lys

  • CAS Number 133605-53-5

  • Empirical Formula (Hill Notation) C84H120N28O27

  • Molecular Weight 1954.02

  •  MDL number MFCD00133077

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... APP(351)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?