• USA Home
  • A4425 - Arachidonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
A4425 Sigma-Aldrich

Arachidonic acid sodium salt

from porcine liver, ≥90% (GC), waxy solid

Synonym: 5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Sodium arachidonate, cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid sodium salt, 20:4

  • CAS Number 6610-25-9

  • Empirical Formula (Hill Notation) C20H31O2Na

  • Molecular Weight 326.45

  •  MDL number MFCD00065458

  •  PubChem Substance ID 24890863

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Lipids, Omega 6 fatty acids and derivatives,
biological source   porcine liver
assay   ≥90% (GC)
form   waxy solid
solubility   methanol: 50 mg/mL, clear, colorless to faintly yellow
storage temp.   −20°C
SMILES string   [Na+].CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC([O-])=O
InChI   1S/C20H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22;/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22);/q;+1/p-1/b7-6-,10-9-,13-12-,16-15-;
InChI key   DDMGAAYEUNWXSI-XVSDJDOKSA-M

Description

Caution

The sodium salts of arachidonic acid are very sensitive to oxidation and will turn yellow and deteriorate rapidly in air. They are packaged under vacuum in sealed ampules. Once opened the product must be used quickly or transferred into an inert atmosphere (dry argon) as soon as possible.

Safety & Documentation

Safety Information

Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
JX3874000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?