• USA Home
  • A4659 - Adenosine 5′-monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
A4659 Sigma-Aldrich

Adenosine 5′-monophosphate sodium salt

from equine muscle, crystalline, suitable for cell culture

Synonym: 5′-Adenylic acid, A-5′-P, AMP

  • CAS Number 149022-20-8

  • Empirical Formula (Hill Notation) C10H14N5O7P · xNa+ · yH2O

  • Molecular Weight 347.22 (anhydrous free acid basis)

  •  PubChem Substance ID 24890891

Purchase

Properties

Related Categories Cell Culture, Miscellaneous Reagents and Supplements, Reagents and Supplements More...
biological source   equine muscle
assay   99-100%
form   crystalline
application(s)   cell culture | mammalian: suitable
storage temp.   −20°C
SMILES string   NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@]((COP(O)(O)=O)([H])O3.C.
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
WGK 3
RTECS 
AU7490000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A1752 Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?