EMAIL THIS PAGE TO A FRIEND
A4876 Sigma-Aldrich

N-Acetyl-DL-proline

Sigma Grade

  • CAS Number 1074-79-9

  • Empirical Formula (Hill Notation) C7H11NO3

  • Molecular Weight 157.17

  •  MDL number MFCD00063230

  •  PubChem Substance ID 24890920

Purchase

Properties

grade   Sigma Grade
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?