EMAIL THIS PAGE TO A FRIEND
A4908 Sigma-Aldrich

Arg-Phe acetate salt

  • CAS Number 102029-92-5

  • Empirical Formula (Hill Notation) C15H23N5O3

  • Molecular Weight 321.37

  •  MDL number MFCD00037260

  •  PubChem Substance ID 24890922

  •  NACRES NA.26

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   JYCGKDWKVVHIHV-UHFFFAOYSA-N
storage temp.   −20°C

Description

Application

L-Arginine-L-Phenyalanine (Arg-Phe) is used as a model compound, aggregation accelerator, in physicochemical analysis of the formation mechanisms and structures of protein aggregates.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?