• USA Home
  • A4916 - (R)-(−)-2-Amino-1-propanol

EMAIL THIS PAGE TO A FRIEND
A4916 Sigma-Aldrich

(R)-(−)-2-Amino-1-propanol

~97%

Synonym: D-Alaninol

  • CAS Number 35320-23-1

  • Linear Formula CH3CH(NH2)CH2OH

  • Molecular Weight 75.11

  •  Beilstein/REAXYS Number 1718866

  •  MDL number MFCD00064413

Purchase

Properties

assay   ~97%
refractive index   n20/D 1.4493 (lit.)
bp   166-168 °C (lit.)
density   0.963 g/mL at 20 °C (lit.)
SMILES string   C[C@@H](N)CO
InChI   1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3/t3-/m1/s1
InChI key   BKMMTJMQCTUHRP-GSVOUGTGSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
3
Flash Point(F) 
181.4 °F
Flash Point(C) 
83 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?