• USA Home
  • A5205 - N-Acetyl-Leu-Val-Phe-al

EMAIL THIS PAGE TO A FRIEND
A5205 Sigma-Aldrich

N-Acetyl-Leu-Val-Phe-al

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)CC(NC(C)=O)C(=O)NC(C(C)C)C(=O)NC(Cc1ccccc1)C=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?