• USA Home
  • A5250 - Nα-Acetyl-D-asparagine

EMAIL THIS PAGE TO A FRIEND
A5250 Sigma-Aldrich

Nα-Acetyl-D-asparagine

~99%

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
assay   ~99%
storage temp.   2-8°C
SMILES string   CC(=O)N[C@H](CC(N)=O)C(O)=O
InChI   1S/C6H10N2O4/c1-3(9)8-4(6(11)12)2-5(7)10/h4H,2H2,1H3,(H2,7,10)(H,8,9)(H,11,12)/t4-/m1/s1
InChI key   HXFOXFJUNFFYMO-SCSAIBSYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?