• USA Home
  • A5425 - γ-Aminobutyric acid-carboxy-14C

EMAIL THIS PAGE TO A FRIEND
A5425 Sigma-Aldrich

γ-Aminobutyric acid-carboxy-14C

Synonym: GABA

  • CAS Number 763-94-0

  • Linear Formula H2N[CH2]314COOH

  • Molecular Weight 103.12

  •  MDL number MFCD00135437

Purchase

Properties

storage temp.   2-8°C
SMILES string   NCCCC(O)=O
Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?