• USA Home
  • A5500 - Nα-Acetyl-L-asparagine

EMAIL THIS PAGE TO A FRIEND
A5500 Sigma-Aldrich

Nα-Acetyl-L-asparagine

~99%

  • CAS Number 4033-40-3

  • Empirical Formula (Hill Notation) C6H10N2O4

  • Molecular Weight 174.15

  •  Beilstein/REAXYS Number 1726196

  •  EC Number 223-716-7

  •  MDL number MFCD00066023

Purchase

Properties

assay   ~99%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?