• USA Home
  • A5507 - 5β-Androstane-3,17-dione

EMAIL THIS PAGE TO A FRIEND
A5507 Sigma-Aldrich

5β-Androstane-3,17-dione

Synonym: Etiocholane-3,17-dione

  • CAS Number 1229-12-5

  • Empirical Formula (Hill Notation) C19H28O2

  • Molecular Weight 288.42

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?