• USA Home
  • A5582 - N-Acetyl-Pen-Arg-Gly-Asp-Cys [Disulfide bridge: 1–5]

EMAIL THIS PAGE TO A FRIEND
A5582 Sigma-Aldrich

N-Acetyl-Pen-Arg-Gly-Asp-Cys [Disulfide bridge: 1–5]

≥97% (HPLC)

  • CAS Number 151171-08-3

  • Empirical Formula (Hill Notation) C22H36N8O9S2

  • Molecular Weight 620.70

  •  MDL number MFCD00237417

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Fibrinogen receptor antagonist; potent inhibitor of blood platelet aggregation.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?