• USA Home
  • A5586 - Arachidonic acid-carboxy-14C

EMAIL THIS PAGE TO A FRIEND
A5586 Sigma-Aldrich

Arachidonic acid-carboxy-14C

≥98% (Radiochemical Purity), ethanol solution

  • CAS Number 3435-81-2

  • Empirical Formula (Hill Notation) C20H32O2

  • Molecular Weight 304.47

  •  MDL number MFCD00189526

Purchase

Properties

assay   ≥98% (Radiochemical Purity)
form   ethanol solution
extent of labeling   40-60 mCi per mmol
mass shift   M+2
storage temp.   −20°C
SMILES string   CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCC[14C](O)=O
Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 3
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?