• USA Home
  • A5651 - 5β-Androstane-3α,17β-diol-11-one

EMAIL THIS PAGE TO A FRIEND
A5651 Sigma-Aldrich

5β-Androstane-3α,17β-diol-11-one

Synonym: 3α,17β-Dihydroxy-5β-androstan-11-one

  • CAS Number 1158-94-7

  • Empirical Formula (Hill Notation) C19H30O3

  • Molecular Weight 306.44

  •  MDL number MFCD00051124

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
SMILES string   CC12CC[C@@H](O)CC1CCC3C4CCC(O)C4(C)CC(=O)C23
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?