• USA Home
  • A5678 - N-Acetyl-D-penicillamine

EMAIL THIS PAGE TO A FRIEND
A5678 Sigma-Aldrich

N-Acetyl-D-penicillamine

≥99%

Synonym: N-Acetyl-3-mercapto-D-valine

  • CAS Number 15537-71-0

  • Empirical Formula (Hill Notation) C7H13NO3S

  • Molecular Weight 191.25

  •  Beilstein/REAXYS Number 1724742

  •  EC Number 239-585-4

  •  MDL number MFCD00078887

Purchase

Properties

InChI Key   MNNBCKASUFBXCO-YFKPBYRVSA-N
assay   ≥99%
storage temp.   2-8°C

Description

Application

Chiral reagent for precolumn derivatization of enantiomeric amino acids or amino alcohols

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?