• USA Home
  • A5909 - N-Acetylprocainamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
A5909 Supelco

N-Acetylprocainamide hydrochloride

~97% (balance primarily procainamide)

Synonym: N-Acetylnovocainamide hydrochloride, Acecainide hydrochloride, NAPA

Purchase

Properties

assay   ~97% (balance primarily procainamide)
mp   184-186 °C (lit.)
storage temp.   −20°C
SMILES string   Cl[H].CCN(CC)CCNC(=O)c1ccc(NC(C)=O)cc1
InChI   1S/C15H23N3O2.ClH/c1-4-18(5-2)11-10-16-15(20)13-6-8-14(9-7-13)17-12(3)19;/h6-9H,4-5,10-11H2,1-3H3,(H,16,20)(H,17,19);1H
InChI key   IYEWBJUCJHKLHD-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Class III antiarrhythmic. Increases the duration of the action potential by decreasing the delayed outward potassium current, slightly decreasing the calcium current, and slightly depressing the inward rectifier potassium current. This is the active metabolite of procainamide that does not induce systemic lupus erythematosus.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?