• USA Home
  • A6259 - N-Acetyl-D-lactosamine

EMAIL THIS PAGE TO A FRIEND
A6259 Sigma-Aldrich

N-Acetyl-D-lactosamine

95-98%

Synonym: β-D-Gal-(1→4)-D-GlcNAc, 2-Acetamido-2-deoxy-4-O-β-D-galactopyranosyl-D-glucopyranose, LN, N-Acetyl-4-O-(β-D-galactopyranosyl)-D-glucosamine

  • CAS Number 32181-59-2

  • Empirical Formula (Hill Notation) C14H25NO11

  • Molecular Weight 383.35

  •  Beilstein/REAXYS Number 96808

  •  MDL number MFCD00063785

Purchase

Properties

assay   95-98%
storage temp.   −20°C
SMILES string   CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H]1O
InChI   1S/C14H25NO11/c1-4(18)15-7-9(20)12(6(3-17)24-13(7)23)26-14-11(22)10(21)8(19)5(2-16)25-14/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5-,6-,7-,8+,9-,10+,11-,12-,13?,14+/m1/s1
InChI key   KFEUJDWYNGMDBV-RPHKZZMBSA-N

Description

Application

Useful in studies of galactosidase, fucosyltransferase, sialyltransferase, and lectin inhibition.

Preparation Note

Enzymatically prepared.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?