• USA Home
  • A6543 - Anthracene Chrome Red A

EMAIL THIS PAGE TO A FRIEND
A6543 Sigma-Aldrich

Anthracene Chrome Red A

~90%

Synonym: Mordant Red 5

  • CAS Number 3564-26-9

  • Linear Formula C12H9N2O6SNa

  • Molecular Weight 332.26

  •  Colour Index Number 14290

  •  EC Number 222-644-3

  •  MDL number MFCD00056115

Purchase

Properties

assay   ~90%
SMILES string   [Na].Oc1ccc(\N=N\c2cc(ccc2O)S(O)(=O)=O)c(O)c1
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?