• USA Home
  • A6651 - 3β-Hydroxy-5α-androstane-17β-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
A6651 Sigma

3β-Hydroxy-5α-androstane-17β-carboxylic acid

Synonym: 3β-Hydroxy-5α-etianic acid

  • CAS Number 15173-54-3

  • Empirical Formula (Hill Notation) C20H32O3

  • Molecular Weight 320.47

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?