• USA Home
  • A6794 - (2S,3R)-3-Amino-2-hydroxy-5-methylhexanoic acid hydrochloride

EMAIL THIS PAGE TO A FRIEND
A6794 Sigma-Aldrich

(2S,3R)-3-Amino-2-hydroxy-5-methylhexanoic acid hydrochloride

  • CAS Number 70853-11-1

  • Empirical Formula (Hill Notation) C7H15NO3 · HCl

  • Molecular Weight 197.66

  •  MDL number MFCD00058431

Purchase

Properties

storage temp.   −20°C

Description

Application

Novel amino acid for the preparation of amastatin and analogs.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?