EMAIL THIS PAGE TO A FRIEND
A6810 Sigma-Aldrich

Acetylsalicylic acid

BioReagent, suitable for plant cell culture, ≥99.0%

Synonym: 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

  • CAS Number 50-78-2

  • Linear Formula 2-(CH3CO2)C6H4CO2H

  • Molecular Weight 180.16

  •  Beilstein/REAXYS Number 779271

  •  EC Number 200-064-1

  •  MDL number MFCD00002430

  •  PubChem Substance ID 24891140

  •  NACRES NA.71

Purchase

Properties

Quality Level   200
product line   BioReagent
assay   ≥99.0%
form   powder
application(s)   cell culture | plant: suitable
mp   134-136 °C (lit.)
solubility   ethanol: 50 mg/mL
SMILES string   CC(=O)Oc1ccccc1C(O)=O
InChI   1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)
InChI key   BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Gene Information   human ... ITGA2B(3674), PTGS1(5742), PTGS2(5743)

Description

Packaging

250 g in poly bottle

Application

Useful for the study of programmed cell death in plants.

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
VO0700000
Flash Point(F) 
482 °F
Flash Point(C) 
250 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?