• USA Home
  • A7397 - 2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose

EMAIL THIS PAGE TO A FRIEND
A7397 Sigma-Aldrich

2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose

≥98%

Synonym: N-Acetyl-D-glucosylamine

  • CAS Number 4229-38-3

  • Empirical Formula (Hill Notation) C8H16N2O5

  • Molecular Weight 220.22

  •  MDL number MFCD00057751

Purchase

Properties

assay   ≥98%
storage temp.   −20°C
SMILES string   CC(=O)NC1C(N)OC(CO)C(O)C1O
InChI   1S/C8H16N2O5/c1-3(12)10-5-7(14)6(13)4(2-11)15-8(5)9/h4-8,11,13-14H,2,9H2,1H3,(H,10,12)
InChI key   MCGXOCXFFNKASF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?