EMAIL THIS PAGE TO A FRIEND
A7519 Sigma-Aldrich

Acetone-d6

Synonym: Hexadeuteroacetone

  • CAS Number 666-52-4

  • Linear Formula CD3COCD3

  • Molecular Weight 64.12

  •  Beilstein/REAXYS Number 1702935

  •  EC Number 211-563-9

  •  MDL number MFCD00044635

Purchase

Properties

vapor density   2 (vs air)
vapor pressure   14.39 psi ( 55 °C)
  184 mmHg ( 20 °C)
  3.67 psi ( 20 °C)
InChI Key   CSCPPACGZOOCGX-WFGJKAKNSA-N
isotopic purity   99.5 atom % D
expl. lim.   13.2 %
refractive index   n20/D 1.355 (lit.)
bp   55.5 °C (lit.)
mp   −93.8 °C (lit.)
density   0.872 g/mL at 25 °C (lit.)
mass shift   M+6
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1090 - class 3 - PG 2 - Acetone
WGK Germany 
3
Flash Point(F) 
-2.2 °F
Flash Point(C) 
-19 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?