• USA Home
  • A7647 - 1,5-Anhydro-D-mannitol

EMAIL THIS PAGE TO A FRIEND
A7647 Sigma-Aldrich

1,5-Anhydro-D-mannitol

Synonym: Styracitol

  • CAS Number 492-93-3

  • Empirical Formula (Hill Notation) C6H12O5

  • Molecular Weight 164.16

  •  MDL number MFCD00067387

Purchase

Properties

SMILES string   OC[C@H]1OC[C@@H](O)[C@@H](O)[C@@H]1O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?