EMAIL THIS PAGE TO A FRIEND
A7660 Sigma-Aldrich

Asp-Phe

Synonym: L-Aspartyl-L-phenylalanine, Aspartame acid

  • CAS Number 13433-09-5

  • Linear Formula HOOCCH2CH(NH2)CONHCH(CH2C6H5)CO2H

  • Molecular Weight 280.28

  •  Beilstein/REAXYS Number 2817528

  •  EC Number 236-557-3

  •  MDL number MFCD00063155

Purchase

Properties

InChI Key   YZQCXOFQZKCETR-UWVGGRQHSA-N
mp   236-239 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

421669 Asp-Phe, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?