• USA Home
  • A8046 - Asparagine-7-amido-4-methylcoumarin trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
A8046 Sigma-Aldrich

Asparagine-7-amido-4-methylcoumarin trifluoroacetate salt

  • CAS Number 115047-90-0

  • Empirical Formula (Hill Notation) C14H15N3O4 · C2HF3O2

  • Molecular Weight 403.31

  •  MDL number MFCD00078916

Purchase

Properties

storage temp.   −20°C
SMILES string   OC(=O)C(F)(F)F.CC1=CC(=O)Oc2cc(NC(=O)[C@@H](N)CC(N)=O)ccc12
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?