• USA Home
  • A8222 - Aluminum hydroxide Gel

EMAIL THIS PAGE TO A FRIEND
A8222 Sigma-Aldrich

Aluminum hydroxide Gel

colloidal suspension

Synonym: Alhydrogel

  • Linear Formula Al(OH)3

  • Molecular Weight 78.00

  •  MDL number MFCD00003420

  •  PubChem Substance ID 24891215

Purchase

Properties

form   colloidal suspension
SMILES string   O[Al](O)O
InChI   1S/Al.3H2O/h;3*1H2/q+3;;;/p-3
InChI key   WNROFYMDJYEPJX-UHFFFAOYSA-K

Description

General description

Alhydrogel is a stable gel which has a uniformly high adsorption capacity and which can be repeatedly autoclaved without losing any of this adsoprtion capacity. As the product itself is a poor heat conductor, care is necessary during autoclaving and constant stirring is advised.

Application

Adjuvant in immunizations

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
What liquid isProduct A8222, Aluminum hydroxide Gel, suspended in?
This is a colloidal suspension in water.
How do I use Product A8222, Aluminum hydroxide Gel?
A depot effect can be accomplished by injection using aluminum hydroxide.The immunogen, either absorbed to the preformed salt, or trapped during precipitation, can be mixed with heat-killed B. pertussis to provide non-specific stimulation of the immune system.Approximately 50-200 μg of protein antigen can be bound by 1 mg aluminum hydroxide. Injections can be done in normal saline. If the antigen is abundant, a titration using a constant amount of antigen versus different amounts of the aluminum hydroxide can be tested. The immunogen/antigen mixture is incubated at room temperature for 20 minutes and then centrifuged at 10,000 g for 10 min.It is recommended to test the supernatant for the presence of antigen to be certain it has been bound by the adjuvant.The samples are now ready for injection. For a review of commonly used adjuvants, see: Harlow, E., and Lane, D. Antibodies: A Laboratory Manual, (1988) which available for purchase as Product number A2926.
Is Product A8222, Aluminum hydroxide Gel, a GMP product?
Alhydrogel is a GMP product. The vendor has been quality audited by producers of human vaccines from the USA. The product has a FDA master file for bulk starting material. The vendor's product code is MF-2143.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?