EMAIL THIS PAGE TO A FRIEND
A8265 Sigma-Aldrich

12-Azidooleic acid

  • CAS Number 57818-47-0

  • Empirical Formula (Hill Notation) C18H33N3O2

  • Molecular Weight 323.47

Purchase

Properties

storage temp.   −20°C

Description

Application

Photosensitive chemical for studies of phospholipid-protein interactions in biological membranes

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?