• USA Home
  • A8584 - 3′-Azido-3′-deoxythymidine-methyl-3H

EMAIL THIS PAGE TO A FRIEND
A8584 Sigma-Aldrich

3′-Azido-3′-deoxythymidine-methyl-3H

≥95% radiochemical purity basis, aqueous solution

Synonym: AZT, Azidothymidine

Purchase

Properties

assay   ≥95% radiochemical purity basis
form   aqueous solution
extent of labeling   1-3 Ci/mmol
storage temp.   2-8°C

Description

Packaging

serum bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?