• USA Home
  • A8677 - Amyloid β-Protein Fragment 12-28

EMAIL THIS PAGE TO A FRIEND
A8677 Sigma-Aldrich

Amyloid β-Protein Fragment 12-28

≥97% (HPLC)

  • CAS Number 107015-83-8

  • Empirical Formula (Hill Notation) C89H135N25O25

  • Molecular Weight 1955.18

  •  MDL number MFCD00133075

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?