• USA Home
  • A8752 - L-Alanine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
A8752 Sigma-Aldrich

L-Alanine methyl ester hydrochloride

  • CAS Number 2491-20-5

  • Linear Formula CH3CH(NH2)COOCH3 · HCl

  • Molecular Weight 139.58

  •  Beilstein/REAXYS Number 3594033

  •  EC Number 219-652-4

  •  MDL number MFCD00063663

Purchase

Properties

InChI Key   IYUKFAFDFHZKPI-DFWYDOINSA-N
mp   109-111 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

330639 L-Alanine methyl ester hydrochloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?