EMAIL THIS PAGE TO A FRIEND
A8802 Sigma-Aldrich

Antide

≥97% (HPLC)

  • CAS Number 112568-12-4

  • Empirical Formula (Hill Notation) C82H108ClN17O14

  • Molecular Weight 1591.29

  •  MDL number MFCD00133104

  •  PubChem Substance ID 24891367

  •  NACRES NA.32

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, LH-RH, Neuropeptides, Peptides and Proteins,
Quality Level   100
assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)CC(NC(=O)C(CCCCNC(=O)c1cccnc1)NC(=O)C(CCCCNC(=O)c2cccnc2)NC(=O)C(CO)NC(=O)C(Cc3cccnc3)NC(=O)C(Cc4ccc(Cl)cc4)NC(=O)C(Cc5ccc6ccccc6c5)NC(C)=O)C(=O)NC(CCCCNC(C)C)C(=O)N7CCCC7C(=O)NC(C)C(N)=O
InChI   1S/C82H108ClN17O14/c1-50(2)41-65(76(108)95-64(26-11-12-37-88-51(3)4)82(114)100-40-18-27-70(100)81(113)91-52(5)71(84)103)96-75(107)63(25-10-14-39-90-73(105)60-23-17-36-87-48-60)93-74(106)62(24-9-13-38-89-72(104)59-22-16-35-86-47-59)94-80(112)69(49-101)99-79(111)68(45-56-19-15-34-85-46-56)98-78(110)67(43-54-29-32-61(83)33-30-54)97-77(109)66(92-53(6)102)44-55-28-31-57-20-7-8-21-58(57)42-55/h7-8,15-17,19-23,28-36,42,46-48,50-52,62-70,88,101H,9-14,18,24-27,37-41,43-45,49H2,1-6H3,(H2,84,103)(H,89,104)(H,90,105)(H,91,113)(H,92,102)(H,93,106)(H,94,112)(H,95,108)(H,96,107)(H,97,109)(H,98,110)(H,99,111)
InChI key   QRYFGTULTGLGHU-UHFFFAOYSA-N
Gene Information   rat ... Gnrhr(81668)

Description

Amino Acid Sequence

Ac-Ala-Phe-Ala-Ser-Lys-Lys-Leu-Lys-Pro-Ala-NH2

Biochem/physiol Actions

LH-RH antagonist with high anti-ovulatory and negligible histamine release activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
AY2961700
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?