• USA Home
  • A8900 - 2-Acetamido-2-deoxy-D-allose

EMAIL THIS PAGE TO A FRIEND
A8900 Sigma

2-Acetamido-2-deoxy-D-allose

  • CAS Number 20880-45-9

  • Empirical Formula (Hill Notation) C8H15NO6

  • Molecular Weight 221.21

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?