EMAIL THIS PAGE TO A FRIEND
A8905 Sigma

15-Acetoxyscirpenol

Synonym: 15-Acetoxyscirpendiol, 15-Acetoxy-3α,4β-dihydroxy-12,13-epoxytrichothec-9-ene

  • CAS Number 2623-22-5

  • Empirical Formula (Hill Notation) C17H24O6

  • Molecular Weight 324.37

  •  MDL number MFCD00078774

  •  PubChem Substance ID 24891379

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
YD0111000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?