• USA Home
  • A9142 - 5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 3′:5′-cyclic-monophosphate,

EMAIL THIS PAGE TO A FRIEND
A9142 Sigma-Aldrich

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 3′:5′-cyclic-monophosphate,

≥90%

Synonym: AICAR 3′:5′-monophosphate, N1-(β-D-3′:5′-Monophosphoribofuranosyl)-5-aminoimidazole-4-carboxamide

Purchase

Properties

assay   ≥90%
storage temp.   −20°C
SMILES string   NC(=O)c1ncn(C2OC3COP(O)(=O)OC3C2O)c1N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?