• USA Home
  • A9201 - N-Acetyl-S-farnesyl-L-cysteine

EMAIL THIS PAGE TO A FRIEND
A9201 Sigma-Aldrich

N-Acetyl-S-farnesyl-L-cysteine

Synonym: AFC

  • CAS Number 135304-07-3

  • Empirical Formula (Hill Notation) C20H33NO3S

  • Molecular Weight 367.55

  •  MDL number MFCD00871540

Purchase

Properties

storage temp.   −20°C

Description

Biochem/physiol Actions

Has high affinity for S-farnesylcysteine methyl transferase thereby inhibiting carboxyterminal methylation of proteins, such as p21ras and Rap1

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?