• USA Home
  • A9638 - 4-Aminophenyl α-L-fucopyranoside

EMAIL THIS PAGE TO A FRIEND
A9638 Sigma

4-Aminophenyl α-L-fucopyranoside

  • CAS Number 42935-25-1

  • Empirical Formula (Hill Notation) C12H17NO5

  • Molecular Weight 255.27

  •  MDL number MFCD00056074

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?