• USA Home
  • A9885 - Nα-Acetyl-L-lysine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
A9885 Sigma-Aldrich

Nα-Acetyl-L-lysine methyl ester hydrochloride

Purchase

Properties

mp   108-114 °C (lit.)
storage temp.   −20°C
SMILES string   Cl.COC(=O)[C@H](CCCCN)NC(C)=O
InChI   1S/C9H18N2O3.ClH/c1-7(12)11-8(9(13)14-2)5-3-4-6-10;/h8H,3-6,10H2,1-2H3,(H,11,12);1H/t8-;/m0./s1
InChI key   PCHGTXYLEAQWOM-QRPNPIFTSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

859095 Nα-Acetyl-L-lysine methyl ester hydrochloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?