• USA Home
  • B001 - Clonidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
B001 Supelco

Clonidine hydrochloride

Synonym: 2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride

  • CAS Number 4205-91-8

  • Empirical Formula (Hill Notation) C9H9Cl2N3 · HCl

  • Molecular Weight 266.55

  •  Beilstein/REAXYS Number 4163525

  •  EC Number 224-121-5

  •  MDL number MFCD00036705

  •  PubChem Substance ID 329773090

Purchase

Properties

form   neat
format   neat
SMILES string   Cl[H].Clc1cccc(Cl)c1NC2=NCCN2
InChI   1S/C9H9Cl2N3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3H,4-5H2,(H2,12,13,14);1H
InChI key   ZNIFSRGNXRYGHF-UHFFFAOYSA-N
Gene Information   human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811PIH 6.1 / PGI
WGK Germany 
3
RTECS 
NJ2490000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C7897 Clonidine hydrochloride, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?