• USA Home
  • B0402 - O-Benzyl-L-threonine hydrochloride

EMAIL THIS PAGE TO A FRIEND
B0402 Sigma-Aldrich

O-Benzyl-L-threonine hydrochloride

  • CAS Number 4378-10-3

  • Empirical Formula (Hill Notation) C11H15NO3 · HCl

  • Molecular Weight 245.70

  •  MDL number MFCD00235875

  •  PubChem Substance ID 24891518

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?