• USA Home
  • B0522 - 5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

EMAIL THIS PAGE TO A FRIEND
B0522 Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

≥98%

Synonym: X-GlcA, X-glucuronide CHA salt

  • CAS Number 114162-64-0

  • Empirical Formula (Hill Notation) C14H13BrClNO7 · C6H13N

  • Molecular Weight 521.79

  •  MDL number MFCD00058543

  •  PubChem Substance ID 24891524

Purchase

Properties

assay   ≥98%
storage temp.   −20°C
SMILES string   NC1CCCCC1.O[C@@H]2[C@@H](O)[C@@H](O[C@@H]([C@H]2O)C(O)=O)Oc3c[nH]c4ccc(Br)c(Cl)c34
InChI   1S/C14H13BrClNO7.C6H13N/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1
InChI key   JXCKZXHCJOVIAV-CYRSAHDMSA-N

Description

Application

Chromogenic substrate for β-glucuronidase (GUS) gene detection.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Fermentation: Enzymes, Substrates and SCFA

The influence of probiotics on the metabolic activity of intestinal microbiota may be evaulated by detecting bacterial enzymatic activity in fecal samples, specifically β-glucosidase, β-glucuronidase...
Chloe McClanahan
Biofiles Edition 6.2
Keywords: Absorption, Cancer, Carcinogens, Cell proliferation, Chromatography, Enzyme activity, Gas chromatography, Gastrointestinal, High performance liquid chromatography, Metabolites, Prebiotics, Probiotics, Thin layer chromatography

Probiotics and Human Health

At the beginning of the 20th century, probiotics were thought to have a beneficial effect to the host by improving the intestinal microbial balance. Through his studies and observations, the Russian ...
Chloe McClanahan
BioFiles Edition 6.2
Keywords: AGE, Absorption, Adhesion, Amplification, Anaerobic, Antimicrobials, Cancer, Carcinogens, Cell proliferation, Chromatography, Coagulation, Detoxification, Diagnostic, Diseases, Enzyme activity, Fermentation, Gas chromatography, Gastrointestinal, High performance liquid chromatography, Inflammation, Magnetic resonance spectroscopy, Metabolism, Metabolites, Molecular biology, Nucleic acid hybridization, PAGE, Pharmaceutical, Phosphorylations, Polymerase chain reaction, Prebiotics, Probiotics, Reductions, Respiratory, Sequencing, Thin layer chromatography, Titrations

Related Products

related product

Product #

Description

Add to Cart

GUSS β-Glucuronidase Reporter Gene Staining Kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?