EMAIL THIS PAGE TO A FRIEND
B0641 Sigma-Aldrich

Boc-Asp(OcHx)-OH

Synonym: Boc-L-aspartic acid 4-cyclohexyl ester

  • CAS Number 73821-95-1

  • Linear Formula C6H11OCOCH2CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 315.36

  •  Beilstein/REAXYS Number 3563576

  •  MDL number MFCD00061996

Purchase

Properties

InChI Key   NLPQIWFEEKQBBN-NSHDSACASA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15398 Boc-Asp(OcHx)-OH, ≥98.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?