• USA Home
  • B113 - (±)−BMAA Hydrochloride

EMAIL THIS PAGE TO A FRIEND
B113 Sigma-Aldrich

(±)−BMAA Hydrochloride

Purchase

Properties

form   solid
color   white
mp   181-182 °C
solubility   H2O: soluble
SMILES string   Cl.CNCC(N)C(O)=O

Description

Application

Synthetic preparation of a neurotoxic amino acid originally isolated from Cycas circinalis.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UE6825000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?