• USA Home
  • B1517 - Boc-Asp(OtBu)-O-(NHS)

EMAIL THIS PAGE TO A FRIEND
B1517 Sigma-Aldrich

Boc-Asp(OtBu)-O-(NHS)

Synonym: Boc-L-aspartic acid 4-(tert-butyl) 1-(hydroxysuccinimide) ester

  • CAS Number 72671-27-3

  • Empirical Formula (Hill Notation) C17H26N2O8

  • Molecular Weight 386.40

  •  MDL number MFCD00063389

  •  PubChem Substance ID 24891596

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)(C)OC(=O)C[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
InChI   1S/C17H26N2O8/c1-16(2,3)25-13(22)9-10(18-15(24)26-17(4,5)6)14(23)27-19-11(20)7-8-12(19)21/h10H,7-9H2,1-6H3,(H,18,24)/t10-/m0/s1
InChI key   PIITZDSTZQZNQH-JTQLQIEISA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?