• USA Home
  • B159 - (±)-1-(4-Bromo-2,5-dimethoxyphenyl)-2-ethanamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
B159 Sigma-Aldrich

(±)-1-(4-Bromo-2,5-dimethoxyphenyl)-2-ethanamine hydrochloride

solid

Synonym: (±)-4-Bromo-2,5-dimethoxybenzeneethanamine hydrochloride, Nexus

  • CAS Number 66142-81-2

  • Empirical Formula (Hill Notation) C10H14BrNO2 · HCl

  • Molecular Weight 296.59

  •  MDL number MFCD00274029

  •  PubChem Substance ID 24891599

Purchase

Properties

form   solid
drug control   USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
color   white
solubility   H2O: soluble
SMILES string   Cl.COc1cc(CCN)c(OC)cc1Br

Description

Biochem/physiol Actions

Hallucinogen; analog of (±)-DOB HBr (Prod. No. D7412).

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?