EMAIL THIS PAGE TO A FRIEND
B2879 Sigma-Aldrich

N-Benzoyl-β-alanine

  • CAS Number 3440-28-6

  • Empirical Formula (Hill Notation) C10H11NO3

  • Molecular Weight 193.20

  •  MDL number MFCD00037293

  •  PubChem Substance ID 24891413

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
storage temp.   −20°C
SMILES string   OC(=O)CCNC(=O)c1ccccc1
InChI   1S/C10H11NO3/c12-9(13)6-7-11-10(14)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,14)(H,12,13)
InChI key   CWXYHOHYCJXYFQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
AY3272500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?