• USA Home
  • B3144 - Boc-Trp-Met-Phe amide

EMAIL THIS PAGE TO A FRIEND
B3144 Sigma-Aldrich

Boc-Trp-Met-Phe amide

≥97% (TLC)

Synonym: des-Asp3-Gastrin related peptide

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cholecystokinins, Neuropeptides, Peptides and Proteins,
InChI Key   KYRVGMSGNSHJEA-UHFFFAOYSA-N
assay   ≥97% (TLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?