• USA Home
  • B3387 - N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine

EMAIL THIS PAGE TO A FRIEND
B3387 Sigma-Aldrich

N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine

≥97.0%

Synonym: Bz-DMT-dA

  • CAS Number 64325-78-6

  • Empirical Formula (Hill Notation) C38H35N5O6

  • Molecular Weight 657.71

  •  Beilstein/REAXYS Number 602310

  •  EC Number 264-776-4

  •  MDL number MFCD00010058

  •  PubChem Substance ID 24891715

Purchase

Properties

assay   ≥97.0%
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2O)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7
InChI   1S/C38H35N5O6/c1-46-29-17-13-27(14-18-29)38(26-11-7-4-8-12-26,28-15-19-30(47-2)20-16-28)48-22-32-31(44)21-33(49-32)43-24-41-34-35(39-23-40-36(34)43)42-37(45)25-9-5-3-6-10-25/h3-20,23-24,31-33,44H,21-22H2,1-2H3,(H,39,40,42,45)/t31-,32+,33+/m0/s1
InChI key   LPICNYATEWGYHI-WIHCDAFUSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?