• USA Home
  • B3667 - 1,3-Benzodithiolylium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
B3667 Sigma-Aldrich

1,3-Benzodithiolylium tetrafluoroborate

≥97% (elemental analysis)

Synonym: BDTF

  • CAS Number 57842-27-0

  • Empirical Formula (Hill Notation) C7H5BF4S2

  • Molecular Weight 240.05

  •  Beilstein/REAXYS Number 4731453

  •  MDL number MFCD00070613

Purchase

Properties

assay   ≥97% (elemental analysis)
mp   150 °C (dec.) (lit.)
storage temp.   2-8°C
SMILES string   F[B-](F)(F)F.c1ccc2[s+]csc2c1
InChI   1S/C7H5S2.BF4/c1-2-4-7-6(3-1)8-5-9-7;2-1(3,4)5/h1-5H;/q+1;-1
InChI key   CUSWPJQKCZMDPY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?